3,5-Dichlorphenylhydraziniumchlorid

Lieferant: Thermo Scientific

Synonyme: 1-(3,5-Dichlorophenyl)hydrazine hydrochloride, 3,5-Dichlorophenylhydrazine hydrochloride

BTB08665.1 BTB08665.10 BTB08665.50
MAYBBTB08665.1EA 0 EUR
MAYBBTB08665.1 MAYBBTB08665.10 MAYBBTB08665.50
3,5-Dichlorphenylhydraziniumchlorid
3,5-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
MDL Number: MFCD00012938
CAS-Nummer: 63352-99-8
EINECS: 264-109-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR