3,3'-Diaminodiphenylsulfon 98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
3,3'-Sulphonyldianiline, 3,3'-sulphonyldianiline, 3-Aminophenyl sulphone, DADPS
L11400.14
L11400.22
L11400.14EA
34.2
EUR
L11400.14
L11400.22
3,3'-Diaminodiphenylsulfon 98%
3,3'-Diaminodiphenylsulfon
Purity: 98%
CAS No.: 599-61-1
Molecular Formula: C12H12N2O2S
Formula Weight: 248.30
Odour: Odourless
Melting Point: 171-174°
Beilstein Reference: 2215938
MDL No.: MFCD00007792
Formel:
C₁₂H₁₂N₂O₂S Schmelzpunkt: 171…174 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007792 CAS-Nummer: 599-61-1 EINECS: 209-967-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...