Bis(4-chlorphenyl)sulfon ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
4,4'-Dichloro diphenyl sulphone, 4-Chlorophenyl sulphone, 4,4'-sulphonylbis(chlorobenzene)
A18938.22
A18938.36
A18938.22EA
34
EUR
A18938.22
A18938.36
Bis(4-chlorphenyl)sulfon ≥99%
Bis(4-chlorphenyl)sulfon
Purity: 99%
CAS No.: 80-07-9
Molecular Formula: C12H8Cl2O2S
Formula Weight: 287.17
Flash Point: 233°(451°F)
Melting Point: 147-150°
Boiling Point: 249-251°/10mm
Beilstein Reference: 2052955
MDL No.: MFCD00000619
Formel:
C₁₂H₈Cl₂O₂S Siedepunkt: 249…251 °C (10 mmHg) Schmelzpunkt: 147…150 °C Flammpunkt: 233 °C (451 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00000619 CAS-Nummer: 80-07-9 EINECS: 201-247-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...