4-Fluorphenylhydraziniumchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride

A15624.06 A15624.14 A15624.22
A15624.06EA 48.1 EUR
A15624.06 A15624.14 A15624.22
4-Fluorphenylhydraziniumchlorid ≥98%
4-Fluorphenylhydraziniumchlorid

Purity: 98% CAS No.: 823-85-8 Molecular Formula: C6H7FN2.HCl Formula Weight: 162.60 Melting Point: ca 265° dec. Beilstein Reference: 3627721

Formel: C₆H₈ClFN₂
Schmelzpunkt: ∼ 265 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012942
CAS-Nummer: 823-85-8
EINECS: 212-521-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR