Methyl-3,4,5-trimethoxybenzoat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
3,4,5-Trimethoxybenzoic acid methyl ester, Methyl 3,4,5-trimethoxybenzoate, Trimethylgallic Acid Methyl Ester
A11933.14
A11933.22
A11933.36
A11933.14EA
26
EUR
A11933.14
A11933.22
A11933.36
Methyl-3,4,5-trimethoxybenzoat ≥98%
Methyl-3,4,5-trimethoxybenzoat
Purity: 98+%
CAS No.: 1916-07-0
Molecular Formula: C11H14O5
Formula Weight: 226.23
Melting Point: 81-85°
Boiling Point: 274-275°
MDL No.: MFCD00008431
Formel:
C₁₁H₁₄O₅ Siedepunkt: 274…275 °C Schmelzpunkt: 81…85 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008431 CAS-Nummer: 1916-07-0 EINECS: 217-629-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...