Chlordimethylvinylsilan 97%

Lieferant: Thermo Fisher Scientific
Gefahr

336730050 336730250
ACRO336730050EA 29.8 EUR
ACRO336730050 ACRO336730250
Chlordimethylvinylsilan 97%
Chlordimethylvinylsilan
Formel: C₄H₉ClSi
Flammpunkt: –4 °C
MDL Number: MFCD00018090
CAS-Nummer: 1719-58-0
EINECS: 217-007-1
UN-Nummer: 2985
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4170 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR