1-Brom-4-fluorbenzol ≥99%
Lieferant: Thermo Fisher Scientific
A11630.0E A11630.22 A11630.36
A11630.0EEA 537 EUR
A11630.0E A11630.22 A11630.36
1-Brom-4-fluorbenzol ≥99%
1-Brom-4-fluorbenzol
Purity: 99% CAS No.: 460-00-4 Molecular Formula: C6H4BrF Formula Weight: 175.01 Flash Point: 60°(140°F) Refractive Index: 1.5270 Odour: Aromatic Melting Point: -16° Boiling Point: 151-153° Beilstein Reference: 774102 UN No.: 1993 MDL No.: MFCD00000342