1-Brom-2,3-difluorbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,3-Difluorobromobenzene
B21585.03
B21585.06
B21585.03EA
41.5
EUR
B21585.03
B21585.06
1-Brom-2,3-difluorbenzol ≥98%
1-Bromo-2,3-difluorobenzene
Purity: 98%
CAS No.: 38573-88-5
Molecular Formula: C6H3BrF2
Formula Weight: 192.99
Flash Point: 61°(141°F)
Refractive Index: 1.5090
Boiling Point: 69-70°/12mm
MDL No.: MFCD00061136
Formel:
C₆H₃BrF₂ Siedepunkt: 69…70 °C (12 mmHg) Dichte: 1.724 Flammpunkt: 61 °C (141 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00061136 CAS-Nummer: 38573-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...