1-Brom-4-chlor-2-nitrobenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A12635.06
A12635.14
A12635.22
A12635.06EA
29.9
EUR
A12635.06
A12635.14
A12635.22
1-Brom-4-chlor-2-nitrobenzol ≥98%
1-Brom-4-chlor-2-nitrobenzol
Purity: 98%
CAS No.: 41513-04-6
Molecular Formula: C6H3BrClNO2
Formula Weight: 236.45
Melting Point: 68-72°
Beilstein Reference: 1950424
Formel:
C₆H₃BrClNO₂ Schmelzpunkt: 68…72 °C Dichte: 2.048 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00024320 CAS-Nummer: 41513-04-6 EINECS: 255-421-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...