4-Brom-3-chloranilin 96%

Lieferant: Thermo Fisher Scientific
Gefahr

L03268.09 L03268.18
L03268.09EA 79.6 EUR
L03268.09 L03268.18
4-Brom-3-chloranilin 96%
4-Brom-3-chloranilin

Purity: 96% CAS No.: 21402-26-6 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Odour: Characteristic Melting Point: 61-66° Beilstein Reference: 1210160 UN No.: 2811 MDL No.: MFCD00041311

Formel: C₆H₅BrClN
Siedepunkt: 264 °C (1013 hPa)
Schmelzpunkt: 61…66 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00041311
CAS-Nummer: 21402-26-6
EINECS: 244-370-3
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR