5-Bromsalicylaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Bromo-2-hydroxybenzaldehyde

A11917.14 A11917.22 A11917.36
A11917.14EA 37 EUR
A11917.14 A11917.22 A11917.36
5-Bromsalicylaldehyd ≥98%
5-Bromsalicylaldehyd

Purity: 98% CAS No.: 1761-61-1 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Melting Point: 103-106° Beilstein Reference: 774710

Formel: C₇H₅BrO₂
Schmelzpunkt: 103…106 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003330
CAS-Nummer: 1761-61-1
EINECS: 217-167-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR