trans-2-Penten ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (E)-2-Pentene

L09421.06 L09421.03
L09421.06EA 111 EUR
L09421.06 L09421.03
trans-2-Penten ≥99%
trans-2-Penten

Purity: 99% CAS No.: 646-04-8 Molecular Formula: C5H10 Formula Weight: 70.14 Storage Temperature: 2°C to 8°C Flash Point: -45°(-49°F) Refractive Index: 1.3790 Odour: Petroleum-like Melting Point: -136° Boiling Point: 36-37° Beilstein Reference: 1718795 Merck Reference: 14,7123 UN No.: 3295 MDL No.: MFCD00009384

Formel: C₅H₁₀
Siedepunkt: 36…37 °C
Schmelzpunkt: –136 °C
Dichte: 0.649
Flammpunkt: –45 °C (–49 °F)
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00009384
CAS-Nummer: 646-04-8
EINECS: 211-461-4
UN-Nummer: 3295
ADR: 3,II
Merck Index: 14,7123

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR