Isovaleraldehyd ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Methylbutyraldehyde
A19640.AE
A19640.AP
A19640.AU
A19640.AEEA
28.5
EUR
A19640.AE
A19640.AP
A19640.AU
Isovaleraldehyd ≥98%
Isovaleraldehyd
Purity: 98%
CAS No.: 590-86-3
Molecular Formula: (CH3)2CHCH2CHO
Formula Weight: 86.13
Flash Point: -1°(30°F)
Refractive Index: 1.3882
Odour: Fruit-like
Melting Point: -51°
Boiling Point: 91-93°
Beilstein Reference: 773692
Merck Reference: 14,5229
UN No.: 2058
MDL No.: MFCD00007014
Formel:
C₅H₁₀O Siedepunkt: 91…93 °C Schmelzpunkt: –51 °C Dichte: 0.800 Flammpunkt: –1 °C (30 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007014 CAS-Nummer: 590-86-3 EINECS: 209-691-5 UN-Nummer: 2058 ADR: 3,II Merck Index: 14,5229 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...