1-Brom-2-methylpropan ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Isobutyl bromide
Purity: 98+%
CAS No.: 78-77-3
Molecular Formula: C4H9Br
Formula Weight: 137.02
Flash Point: 18°(64°F)
Refractive Index: 1.4350
Melting Point: -118°
Boiling Point: 90-92°
Solubility: Slightly soluble in water. Miscible with alcohol, ether
Beilstein Reference: 1730915
Merck Reference: 14,5135
UN No.: 2342
MDL No.: MFCD00000217
Formel:
C₄H₉Br Molekulargewicht: 137.02 g/mol Siedepunkt: 90…92 °C Schmelzpunkt: –118 °C Dichte: 1.255 Flammpunkt: 18 °C (64 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00000217 CAS-Nummer: 78-77-3 EINECS: 201-141-2 UN-Nummer: 2342 ADR: 3 II Merck Index: 13,05152 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...