ω-Brom-3-hydroxyacetophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
α-Bromo-3-hydroxyacetophenone, 3-Hydroxyphenacyl bromide, 2-Bromo-1-(3-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(3-hydroxyphenyl)ethanone, 2-Bromo-3'-hydroxyacetophenone
M-Clarity™ Qualitätsstufe = MQ100
594709-1G
594709-5G
SIAL594709-1GEA
112
EUR
SIAL594709-1G
SIAL594709-5G
ω-Brom-3-hydroxyacetophenon, Sigma-Aldrich®
ω-Brom-3-hydroxyacetophenon
Formel:
C₈H₇BrO₂ Molekulargewicht: 215.05 |
MDL Number:
MFCD05664326 CAS-Nummer: 2491-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...