Kalium(4-fluorphenyl)trifluoroborat 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Fluorophenyl)trifluoroboric acid potassium salt, Potassium trifluoro(4-fluorophenyl)borate

L17655.03 L17655.06
L17655.03EA 58.7 EUR
L17655.03 L17655.06
Kalium(4-fluorphenyl)trifluoroborat 95%
Kalium(4-fluorphenyl)trifluoroborat

Purity: 95% CAS No.: 192863-35-7 Molecular Formula: C6H4BF4K Formula Weight: 202.00 Melting Point: 290-292° Beilstein Reference: 7783544

Formel: C₆H₄BF₄K
Schmelzpunkt: 290…292 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01318170
CAS-Nummer: 192863-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR