Kalium(4-methylphenyl)trifluoroborat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Potassium trifluoro(p-tolyl)borate, Potassium p-tolyltrifluoroborate, (4-Methylphenyl)trifluoroboric acid potassium salt
L17604.03
L17604.06
L17604.03EA
51.5
EUR
L17604.03
L17604.06
Kalium(4-methylphenyl)trifluoroborat ≥98%
Kalium(4-methylphenyl)trifluoroborat
Purity: 98%
CAS No.: 216434-82-1
Molecular Formula: C7H7BF3K
Formula Weight: 198.04
Melting Point:
Formel:
C₇H₇BF₃K Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01318171 CAS-Nummer: 216434-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...