3-(Trimethylsilyl)propiolsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H53380.03
H53380.06
H53380.14
H53380.03EA
70.2
EUR
H53380.03
H53380.06
H53380.14
3-(Trimethylsilyl)propiolsäure ≥97%
3-(Trimethylsilyl)propiolsäure
Purity: 97%
CAS No.: 5683-31-8
Molecular Formula: C6H10O2Si
Formula Weight: 142.23
Flash Point: 96°(205°F)
Melting Point: 47-49°
Boiling Point: 105-110°/10mm
Beilstein Reference: 1755674
MDL No.: MFCD00190216
Formel:
C₆H₁₀O₂Si Siedepunkt: 105…110 °C (10 mmHg) Schmelzpunkt: 47…49 °C Flammpunkt: 96 °C (205 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00190216 CAS-Nummer: 5683-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...