Natriumoctylsulfat, Pulver

Lieferant: MP Biomedicals

Synonyme: Octyl sulphate, sodium salt, Octyl sulphuric acid salt

0215598025 0215598001 0215598005
ICNA0215598025EA 641 EUR
ICNA0215598025 ICNA0215598001 ICNA0215598005
Natriumoctylsulfat, Pulver
Natriumoctylsulfat

Sodium octyl sulphate is an anionic detergent and ion pairing reagent that is used in HPLC. The anionic sulphonate counterion permits the separation and resolution of positively charged analytes.


Sodium octyl sulphate is an anionic detergent and ion pairing reagent that is used in HPLC. The anionic sulphonate counterion permits the separation and resolution of positively charged analytes.


Easily soluble in water (20 mg/ml).

Formel: C₈H₁₇NaO₄S
Molekulargewicht: 232.28 g/mol
Schmelzpunkt: 181…183 °C
Dichte: 1.266 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007470
CAS-Nummer: 142-31-4
EINECS: 205-535-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR