2-Propanol ≥99,5% (durch GC), EMPLURA®, Supelco®
Lieferant: Merck
Gefahr
Synonyme: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
M-Clarity™ quality level = MQ200
8.18766.1000 8.18766.2500 8.18766.9025 8.18766.9180 8.18766.2500UK 8.18766.9011
8.18766.1000EA 48.54 EUR
8.18766.1000 8.18766.2500 8.18766.9025 8.18766.9180 MERC8.18766.2500UK 8.18766.9011
2-Propanol ≥99,5% (durch GC), EMPLURA®, Supelco®
2-Propanol
EMPLURA® is an alternative to high-purity qualities. EMPLURA® is a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Formel: (CH₃)₂CHOH
Siedepunkt: 82 °C (1013 hPa)
Schmelzpunkt: –89 °C
Dichte: 0.786 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN-Nummer: 1219
ADR: 3,II
REACH-Nummer: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM