Propylamin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Propylamine, 1-Aminopropane

36635.AK 36635.K2
36635.AKEA 41.6 EUR
36635.AK 36635.K2
Propylamin ≥99%
Propylamin

Purity: 99+% CAS No.: 107-10-8 Molecular Formula: CH3CH2CH2NH2 Formula Weight: 59.11 Flash Point: -12°(10°F) Physical Form: Liquid Density: 0.719 Refractive Index: 1.389 Sensitivity: Air Sensitive Appearance: Colourless Odour: Amine-like pH: 12.6 Melting Point: -83° Boiling Point: 48-49° Beilstein Reference: 1098243 Merck Reference: 14,7843 UN No.: 1277 MDL No.: MFCD00008205

Formel: H₃CCH₂CH₂NH₂
Molekulargewicht: 59.11 g/mol
Siedepunkt: 48…49 °C
Schmelzpunkt: –83 °C
Dichte: 0.719
Flammpunkt: –12 °C (10 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008205
CAS-Nummer: 107-10-8
EINECS: 203-462-3
UN-Nummer: 1277
ADR: 3 II
Merck Index: 12,08028

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR