1-Chlor-6-iodhexan ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L05198.09 L05198.18
L05198.09EA 102 EUR
L05198.09 L05198.18
1-Chlor-6-iodhexan ≥97%
1-Chlor-6-iodhexan

Purity: 97% CAS No.: 34683-73-3 Molecular Formula: Cl(CH2)6I Formula Weight: 246.52 Storage Temperature: -30°C to -10°C Refractive Index: 1.5250 Beilstein Reference: 1732408 MDL No.: MFCD00039417

Formel: C₆H₁₂ClI
MDL Number: MFCD00039417
CAS-Nummer: 34683-73-3
EINECS: 252-144-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR