Bariumacetat 99,0-102,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Barium di(acetate), Acetic acid barium salt
12198.30
12198.A1
12198.A7
12198.30EA
66.7
EUR
12198.30
12198.A1
12198.A7
Bariumacetat 99,0-102,0% ACS
Bariumacetat
Purity: 99.0-102.0%
CAS No.: 543-80-6
Molecular Formula: Ba(OOCCH3)2
Formula Weight: 255.43
Physical Form: Crystalline
Density: 2.47
Merck Reference: 14,965
UN No.: 1564
MDL No.: MFCD00012447
Formel:
(H₃CCOO)₂Ba Molekulargewicht: 255.42 g/mol Schmelzpunkt: ∼450 °C Dichte: 2.47 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012447 CAS-Nummer: 543-80-6 EINECS: 208-849-0 UN-Nummer: 1564 ADR: 6.1 III Merck Index: 14,965 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...