2,4-Nonandion ≥98%

Lieferant: Thermo Fisher Scientific

L01151.06 L01151.14
L01151.06EA 119 EUR
L01151.06 L01151.14
2,4-Nonandion ≥98%
2,4-Nonandion

Purity: 98+% CAS No.: 6175-23-1 Molecular Formula: C9H16O2 Formula Weight: 156.23 Refractive Index: 1.4608 Boiling Point: 100-103°/19mm MDL No.: MFCD00015276

Formel: C₉H₁₆O₂
Siedepunkt: 100…103 °C (19 mmHg)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00015276
CAS-Nummer: 6175-23-1
EINECS: 228-219-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR