Triethylammoniumdihydrogentrifluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

344648-100G 344648-25G 344648-5G
SIAL344648-100GEA 221 EUR
SIAL344648-100G SIAL344648-25G SIAL344648-5G
Triethylammoniumdihydrogentrifluorid, Sigma-Aldrich®
Triethylammonium dihydrogentrifluoride
Formel: C₆H₁₈F₃N
Dichte: 1,094 g/cm³ (20 °C)
MDL Number: MFCD00043294
CAS-Nummer: 73602-61-6
EINECS: 277-550-5
UN-Nummer: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR