Methylmagnesiumbromid 3 mol/l in Diethylether, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
189898-100ML
189898-20L
189898-50ML
189898-800ML
SIAL189898-100MLEA
52.3
EUR
SIAL189898-100ML
SIAL189898-20L
SIAL189898-50ML
SIAL189898-800ML
Methylmagnesiumbromid 3 mol/l in Diethylether, Sigma-Aldrich®
Methylmagnesium bromide
Formel:
CH₃BrMg Molekulargewicht: 119.24 g/mol |
MDL Number:
MFCD00000041 CAS-Nummer: 75-16-1 EINECS: 200-844-1 UN-Nummer: 1928 ADR: 4.3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...