N-(Hydroxymethyl)benzamid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A12250.09 A12250.14 A12250.30
A12250.09EA 38.5 EUR
A12250.09 A12250.14 A12250.30
N-(Hydroxymethyl)benzamid ≥98%
N-(Hydroxymethyl)benzamid

Purity: 98% CAS No.: 6282-02-6 Molecular Formula: C6H5CONHCH2OH Formula Weight: 151.17 Melting Point: 94-96° Beilstein Reference: 2045129

Formel: C₈H₉NO₂
Molekulargewicht: 151.16 g/mol
Schmelzpunkt: 94…96 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00014415
CAS-Nummer: 6282-02-6
EINECS: 228-495-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR