Bariumhydroxid Monohydrat ≥95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
45888.22
45888.36
45888.22EA
41.6
EUR
45888.22
45888.36
Bariumhydroxid Monohydrat ≥95%
Bariumhydroxid Monohydrat
Purity: 95%
CAS No.: 22326-55-2
Molecular Formula: H2Ba2O2.H2O
Formula Weight: 189.38 (171.36anhy)
Physical Form: Crystalline
Density: 3.743
Appearance: White
Odour: Odourless
UN No.: 3262
MDL No.: MFCD00149151
Formel:
H₂BaO₂·1H₂O Dichte: 3.743 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149151 CAS-Nummer: 22326-55-2 EINECS: 241-234-5 UN-Nummer: 3262 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...