Dexamethason, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
D9184-100MG
D9184-1G
D9184-5G
SIALD9184-100MGEA
194
EUR
SIALD9184-100MG
SIALD9184-1G
SIALD9184-5G
Dexamethason, Sigma-Aldrich®
Dexamethason
Formel:
C₂₂H₂₉FO₅ Schmelzpunkt: 268…271 °C |
MDL Number:
MFCD00064136 CAS-Nummer: 50-02-2 EINECS: 200-003-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...