6-Sulfanyl-1-hexanol, Sigma-Aldrich®

Lieferant: Merck

451088-25ML 451088-5ML
SIAL451088-25MLEA 307 EUR
SIAL451088-25ML SIAL451088-5ML
6-Sulfanyl-1-hexanol, Sigma-Aldrich®
6-Mercapto-1-hexanol
Formel: C₆H₁₄OS
MDL Number: MFCD00068552
CAS-Nummer: 1633-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR