Methylprednisolon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
M0639-100MG
M0639-1G
M0639-250MG
SIALM0639-100MGEA
176
EUR
SIALM0639-100MG
SIALM0639-1G
SIALM0639-250MG
Methylprednisolon, Sigma-Aldrich®
Methylprednisolon
Formel:
C₂₂H₃₀O₅ Schmelzpunkt: 230 °C |
MDL Number:
MFCD00010591 CAS-Nummer: 83-43-2 EINECS: 201-476-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...