4-(Trifluormethyl)benzolsulfonamid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
α,α,α-Trifluoro-p-toluenesulphonamide
B25243.03
B25243.06
B25243.14
B25243.03EA
37
EUR
B25243.03
B25243.06
B25243.14
4-(Trifluormethyl)benzolsulfonamid ≥97%
4-(Trifluormethyl)benzolsulfonamid
Purity: 97%
CAS No.: 830-43-3
Molecular Formula: C7H6F3NO2S
Formula Weight: 225.19
Melting Point: 178-182°
Beilstein Reference: 2695323
Formel:
C₇H₆F₃NO₂S Schmelzpunkt: 178…182 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00159251 CAS-Nummer: 830-43-3 EINECS: 212-596-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...