5-Brom-2-methoxybenzolsulfonylchlorid

Lieferant: Thermo Scientific

BTB04911DA BTB04911EA
MAYBBTB04911DAEA 0 EUR
MAYBBTB04911DA MAYBBTB04911EA
5-Brom-2-methoxybenzolsulfonylchlorid
5-Brom-2-methoxybenzolsulfonylchlorid
Formel: C₇H₆BrClO₃S
Schmelzpunkt: 107…110 °C
MDL Number: MFCD00051768
CAS-Nummer: 23095-05-8
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR