3-Brom-5-nitrosalicylaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Bromo-2-hydroxy-5-nitrobenzaldehyde
L07071.03
L07071.06
L07071.14
L07071.03EA
28.2
EUR
L07071.03
L07071.06
L07071.14
3-Brom-5-nitrosalicylaldehyd ≥98%
3-Brom-5-nitrosalicylaldehyd
Purity: 98%
CAS No.: 16789-84-7
Molecular Formula: C7H4BrNO4
Formula Weight: 246.02
Melting Point: 147-151°
Beilstein Reference: 1213613
Formel:
C₇H₄BrNO₄ Schmelzpunkt: 147…151 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00051833 CAS-Nummer: 16789-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...