3-Amino-9-ethylcarbazol (AEC), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
9-Ethylcarbazol-3-ylamine, 9-Ethyl-9H-carbazol-3-amine, AEC, 50X, AEC, 3-Amino-9-ethylcarbazole
M-Clarity™ quality level = MQ200
A6926-100TAB
A6926-50TAB
A6926-5TAB
SIALA6926-100TABEA
677
EUR
SIALA6926-100TAB
SIALA6926-50TAB
SIALA6926-5TAB
3-Amino-9-ethylcarbazol (AEC), Sigma-Aldrich®
3-Amino-9-ethyl carbazole (AEC)
Formel:
C₁₄H₁₄N₂ Schmelzpunkt: 113…114 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004964 CAS-Nummer: 132-32-1 EINECS: 205-057-7 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...