2-Iodbenzolsulfonamid
Lieferant: Thermo Scientific
NRB00577.250
NRB00577.1
MAYBNRB00577.250EA
0
EUR
MAYBNRB00577.250
MAYBNRB00577.1
2-Iodbenzolsulfonamid
2-Iodbenzolsulfonamid
Formel:
C₆H₆INO₂S |
CAS-Nummer:
53730-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...