2-Ethoxybenzamid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethenzamide

B25455.22 B25455.36
B25455.22EA 28.8 EUR
B25455.22 B25455.36
2-Ethoxybenzamid ≥97%
2-Ethoxybenzamid

Purity: 97% CAS No.: 938-73-8 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: 131-134° Merck Reference: 14,3731 MDL No.: MFCD00007977

Formel: C₉H₁₁NO₂
Siedepunkt: 333 °C (1013 hPa)
Schmelzpunkt: 131…134 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007977
CAS-Nummer: 938-73-8
EINECS: 213-346-4
Merck Index: 14,3731

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR