2-Brombenzimidazol

Lieferant: Thermo Scientific

Synonyme: 2-Bromo-1H-benzimidazole

CC09010DA CC09010EA
MAYBCC09010DAEA 0 EUR
MAYBCC09010DA MAYBCC09010EA
2-Brombenzimidazol
2-Bromobenzimidazole
Formel: C₇H₅BrN₂
MDL Number: MFCD00160009
CAS-Nummer: 54624-57-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR