2,6-Dimethoxybenzamid
Lieferant: Thermo Scientific
BTB07117.250
BTB07117.1
MAYBBTB07117.250EA
0
EUR
MAYBBTB07117.250
MAYBBTB07117.1
2,6-Dimethoxybenzamid
2,6-Dimethoxybenzamid
Formel:
C₉H₁₁NO₃ |
MDL Number:
MFCD00828821 CAS-Nummer: 21864-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...