2-(Brommethyl)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(Bromomethyl)benzeneboronic acid, Dihydroxy(2-(bromomethyl)phenyl)borane, (2-(Bromomethyl)phenyl)metaboric acid, 2-Bromomethylphenylboronic acid
H52467.03
H52467.06
H52467.03EA
106
EUR
H52467.03
H52467.06
2-(Brommethyl)phenylboronsäure ≥97%
2-(Brommethyl)phenylboronsäure
Purity: 97%
CAS No.: 91983-14-1
Molecular Formula: C7H8BBrO2
Formula Weight: 214.85
Melting Point: 145-148°
MDL No.: MFCD01318941
Formel:
C₇H₈BBrO₂ Schmelzpunkt: 145…148 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01318941 CAS-Nummer: 91983-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...