4-Ethyl-2-methoxyphenol ≥98% FCC, FG Kosher, Halal, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Ethyl-2-hydroxyanisole, 1-Hydroxy-2-methoxy-4-ethyl Benzene, 4-Ethylguaiacol

M-Clarity™ quality level = MQ300

W243604-100G-K W243604-1KG-K W243604-5KG-K
SIALW243604-100G-KEA 149 EUR
SIALW243604-100G-K SIALW243604-1KG-K SIALW243604-5KG-K
4-Ethyl-2-methoxyphenol ≥98% FCC, FG Kosher, Halal, Sigma-Aldrich®
4-Ethyl-2-methoxyphenol
Formel: C₉H₁₂O₂
MDL Number: MFCD00038714
CAS-Nummer: 2785-89-9
EINECS: 220-500-4

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Colorless to Light Yellow
Appearance (Form) Liquid
Refractive index at 20 °C 1.525 - 1.530
Infrared spectrum Conforms to Structure
Purity (GC) ≥98.00%
Specific Gravity 1.061 - 1.064
Solubility (Turbidity) Clear
Solubility (Color) 1ml/1ml,95% Ethanol Colorless to Faint Yellow
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Mercury (Hg) ≤1 ppm
Lead (Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR