4'-Nonylacetophenon

Lieferant: Thermo Scientific

BTB09789.1 BTB09789.10
MAYBBTB09789.1EA 0 EUR
MAYBBTB09789.1 MAYBBTB09789.10
4'-Nonylacetophenon
Formel: C₁₇H₂₆O
MDL Number: MFCD00043717
CAS-Nummer: 37593-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR