5-Chlor-4-fluor-2-nitroanilin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Amino-3-chlor-4-fluor-6-nitrobenzene

A13879.06 A13879.14
A13879.06EA 105 EUR
A13879.06 A13879.14
5-Chlor-4-fluor-2-nitroanilin ≥97%
5-Chlor-4-fluor-2-nitroanilin

Purity: 97% CAS No.: 104222-34-6 Molecular Formula: C6H4ClFN2O2 Formula Weight: 190.56 Melting Point: 141-145° Beilstein Reference: 7202993 UN No.: 2811 MDL No.: MFCD00052698

Formel: C₆H₄ClFN₂O₂
Schmelzpunkt: 141…145 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00052698
CAS-Nummer: 104222-34-6
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR