2-Amino-4-bromphenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-2-hydroxyaniline

708356-1G 708356-5G
SIAL708356-1GEA 53.7 EUR
SIAL708356-1G SIAL708356-5G
2-Amino-4-bromphenol, Sigma-Aldrich®
2-Amino-4-bromphenol
Formel: C₆H₆BrNO
MDL Number: MFCD00235171
CAS-Nummer: 40925-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR