3,3'-Diaminobenzophenon 90+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bis(3-aminophenyl)methanone
L09128.03
L09128.06
L09128.03EA
52.5
EUR
L09128.03
L09128.06
3,3'-Diaminobenzophenon 90+%
3,3'-Diaminobenzophenon
Purity: 90+%
CAS No.: 611-79-0
Molecular Formula: C13H12N2O
Formula Weight: 212.25
Melting Point: 148-152°
Boiling Point: 285°/11mm
MDL No.: MFCD00014774
Formel:
C₁₃H₁₂N₂O Siedepunkt: 285 °C (11 mmHg) Schmelzpunkt: 148…152 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014774 CAS-Nummer: 611-79-0 EINECS: 210-281-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...