1,4-Diaminobutan Dihydrochlorid 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,4-Butanediamine dihydrochloride, Tetramethylenediammonium dichloride, Tetramethylendiamine dihydrochloride, Putrescine dihydrochloride

215110050 215110250 215111000
ACRO215110050EA 23.6 EUR
ACRO215110050 ACRO215110250 ACRO215111000
1,4-Diaminobutan Dihydrochlorid 99+%
1,4-Diaminobutan Dihydrochlorid
Formel: C₄H₁₄Cl₂N₂
Schmelzpunkt: 280 °C
Dichte: 1,31 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012526
CAS-Nummer: 333-93-7
EINECS: 206-375-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR