2,3-Diaminophenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydroxy-o-phenylenediamine

M-Clarity™ quality level = MQ100

340618-1G 340618-5G
SIAL340618-1GEA 39.1 EUR
SIAL340618-1G SIAL340618-5G
2,3-Diaminophenol, Sigma-Aldrich®
2,3-Diaminophenol
Formel: C₆H₈N₂O
MDL Number: MFCD00075199
CAS-Nummer: 59649-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR