1,2-Dibenzoylbenzol ≥97%

Lieferant: Thermo Fisher Scientific

L01592.03 L01592.MD
L01592.03EA 68.8 EUR
L01592.03 L01592.MD
1,2-Dibenzoylbenzol ≥97%
1,2-Dibenzoylbenzol

Purity: 97% CAS No.: 1159-86-0 Molecular Formula: C20H14O2 Formula Weight: 286.33 Melting Point: 145-148° Beilstein Reference: 789077

Formel: C₂₀H₁₄O₂
Siedepunkt: 424 °C (1013 hPa)
Schmelzpunkt: 145…148 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003078
CAS-Nummer: 1159-86-0
EINECS: 214-597-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR