ω-Brom-4-chlor-3-nitroacetophenon
Lieferant: Thermo Scientific
Synonyme:
α-Bromo-4-chloro-3-nitroacetophenone, 2-Bromo-4'-chloro-3'-nitroacetophenone, 4-Chloro-3-nitrophenacyl bromide
JP00700DA
JP00700EA
MAYBJP00700DAEA
0
EUR
MAYBJP00700DA
MAYBJP00700EA
ω-Brom-4-chlor-3-nitroacetophenon
ω-Brom-4-chlor-3-nitroacetophenon
Formel:
C₈H₅BrClNO₃ |
CAS-Nummer:
22019-49-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...