1-(3-Aminopropyl)imidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-(3-Aminopropyl)-imidazole, Imidazole-1-propylamine, 1-(3-Aminopropyl)-1H-imidazole
M-Clarity™ quality level = MQ100
272264-250G
272264-50G
SIAL272264-250GEA
173
EUR
SIAL272264-250G
SIAL272264-50G
1-(3-Aminopropyl)imidazol, Sigma-Aldrich®
1-(3-Aminopropyl)imidazole
Formel:
C₆H₁₁N₃ |
MDL Number:
MFCD00009819 CAS-Nummer: 5036-48-6 EINECS: 225-730-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...